Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-4-fluoro-L-phenylalanine, 95%

Chemical Identifiers

Chemical Name or Material N-Boc-4-fluoro-L-phenylalanine
CAS 41153-30-4
Formula Weight 283.3
MDL Number MFCD00079672
Beilstein 4323475
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6137003

  SDS
alfa aesar™
H6137003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6137006

  SDS
alfa aesar™
H6137006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 83° to 86°C
Formula Weight 283.3
MDL Number MFCD00079672
Color White
CAS 41153-30-4
Chemical Name or Material N-Boc-4-fluoro-L-phenylalanine
Assay 95%
Beilstein 4323475
Physical Form Powder
Quantity 1g