missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-4-chloro-L-phenylalanine, 95%

$60.28 - $892.89

Chemical Identifiers

CAS 68090-88-0
Molecular Formula C14H18ClNO4
Molecular Weight (g/mol) 299.751
MDL Number MFCD00057808
InChI Key BETBOAZCLSJOBQ-NSHDSACASA-N
Synonym boc-phe 4-cl-oh, boc-l-4-chlorophenylalanine, boc-4-chloro-l-phenylalanine, boc-l-4-chlorophe, boc-l-phe 4-cl-oh, n-boc-4-chloro-l-phenylalanine, n-tert-butoxycarbonyl-4-chloro-l-phenylalanine, s-n-boc-4-chlorophenylalanine, s-2-tert-butoxycarbonyl amino-3-4-chlorophenyl propanoic acid, boc-p-chloro-phe-oh
PubChem CID 2734499
IUPAC Name (2S)-3-(4-chlorophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NC(CC1=CC=C(C=C1)Cl)C(=O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6128303 View Documents Alfa Aesar™
H6128303
1g Each for $60.28
 
AAH6128306 View Documents Alfa Aesar™
H6128306
5g Each for $223.61
 
AAH6128314 View Documents Alfa Aesar™
H6128314
25g Each for $892.89
 
Specifications

Chemical Identifiers

68090-88-0
299.751
BETBOAZCLSJOBQ-NSHDSACASA-N
2734499
CC(C)(C)OC(=O)NC(CC1=CC=C(C=C1)Cl)C(=O)O
C14H18ClNO4
MFCD00057808
boc-phe 4-cl-oh, boc-l-4-chlorophenylalanine, boc-4-chloro-l-phenylalanine, boc-l-4-chlorophe, boc-l-phe 4-cl-oh, n-boc-4-chloro-l-phenylalanine, n-tert-butoxycarbonyl-4-chloro-l-phenylalanine, s-n-boc-4-chlorophenylalanine, s-2-tert-butoxycarbonyl amino-3-4-chlorophenyl propanoic acid, boc-p-chloro-phe-oh
(2S)-3-(4-chlorophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid

Specifications

~110°C
68090-88-0
MFCD00057808
boc-phe 4-cl-oh, boc-l-4-chlorophenylalanine, boc-4-chloro-l-phenylalanine, boc-l-4-chlorophe, boc-l-phe 4-cl-oh, n-boc-4-chloro-l-phenylalanine, n-tert-butoxycarbonyl-4-chloro-l-phenylalanine, s-n-boc-4-chlorophenylalanine, s-2-tert-butoxycarbonyl amino-3-4-chlorophenyl propanoic acid, boc-p-chloro-phe-oh
CC(C)(C)OC(=O)NC(CC1=CC=C(C=C1)Cl)C(=O)O
299.751
299.75
N-Boc-4-chloro-L-phenylalanine
1g
C14H18ClNO4
4261946
BETBOAZCLSJOBQ-NSHDSACASA-N
(2S)-3-(4-chlorophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
2734499
95%