missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-4-bromoaniline, 97%

Manufacturer:  Alfa Aesar™ H5522606

 View more versions of this product

Catalog No. AAH5522606



Specifications

Specifications

N-Boc-4-bromoaniline
103°C to 106°C
131818-17-2
MFCD01006612
VLGPDTPSKUUHKR-UHFFFAOYSA-N
tert-butyl N-(4-bromophenyl)carbamate
2773608
Solid
Brown to White
5g
C11H14BrNO2
4-bromo-n-boc aniline
CC(C)(C)OC(=O)NC1=CC=C(C=C1)Br
272.142
272.14
97%

Chemical Identifiers

131818-17-2
272.142
VLGPDTPSKUUHKR-UHFFFAOYSA-N
2773608
CC(C)(C)OC(=O)NC1=CC=C(C=C1)Br
C11H14BrNO2
MFCD01006612
4-bromo-n-boc aniline
tert-butyl N-(4-bromophenyl)carbamate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents