Please login to your online account to display your discounted pricing

N-BOC-4-bromoaniline, +95%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC398550050

 View more versions of this product

Catalog No. AC398550050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 272.14
Physical Form Powder
Assay ≥95.0%
Quantity 5g
Color Beige
Melting Point 102° to 106°C
CAS Min % 95.0
CAS Max % 100.0
Packaging Glass bottle
Synonym N-(tert-Butoxycarbonyl)-4-bromoaniline
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material N-BOC-4-Bromoaniline
Assay Percent Range 95+%
CAS 131818-17-2
MDL Number MFCD01006612
Linear Formula BrC6H4NHCOOC(CH3)3
Molecular Formula C11H14BrNO2
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

Specifications

Chemical Name or Material N-BOC-4-Bromoaniline
CAS 131818-17-2
CAS Min % 95.0
CAS Max % 100.0
Assay ≥95.0%
Assay Percent Range 95+%
Molecular Formula C11H14BrNO2
Linear Formula BrC6H4NHCOOC(CH3)3
Formula Weight 272.14
MDL Number MFCD01006612
Synonym N-(tert-Butoxycarbonyl)-4-bromoaniline
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

SDS & Safety Documents