Please login to your online account to display your discounted pricing

N-BOC-4-bromoaniline, +95%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material N-BOC-4-Bromoaniline
CAS 131818-17-2
CAS Min % 95.0
CAS Max % 100.0
Assay ≥95.0%
Assay Percent Range 95+%
Molecular Formula C11H14BrNO2
Linear Formula BrC6H4NHCOOC(CH3)3
Formula Weight 272.14
MDL Number MFCD01006612
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC398550050

  SDS
thermo fisher scientific
AC398550050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC398550250

  SDS
thermo fisher scientific
AC398550250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder
Formula Weight 272.14
Assay ≥95.0%
Quantity 5g
Color Beige
Melting Point 102° to 106°C
CAS Min % 95.0
CAS Max % 100.0
Packaging Glass bottle
Synonym N-(tert-Butoxycarbonyl)-4-bromoaniline
Chemical Name or Material N-BOC-4-Bromoaniline
Assay Percent Range 95+%
MDL Number MFCD01006612
CAS 131818-17-2
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C11H14BrNO2
Linear Formula BrC6H4NHCOOC(CH3)3
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Infrared Spectrum Authentic
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.