Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-4,5-dehydro-L-leucine dicyclohexylamine salt, 97%

Chemical Identifiers

Chemical Name or Material N-Boc-4,5-dehydro-L-leucine dicyclohexylamine salt
CAS 87720-54-5
Molecular Formula C11H19NO4·C12H23N
Formula Weight 410.59
MDL Number MFCD00237524
UN Number UN3077
Synonym (S)-2-(Boc-amino)-4-methyl-4-pentenoic acid dicyclohexylamine salt; Boc-4,5-dehydro-Leu-OH.DCHA
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH27005MF

  SDS
alfa aesar™
H27005MF
50mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 410.59
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
Quantity 50mg
Optical Rotation +13° (c=1 in Methanol)
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD00237524
Chemical Name or Material N-Boc-4,5-dehydro-L-leucine dicyclohexylamine salt
Assay 97%
CAS 87720-54-5
Molecular Formula C11H19NO4·C12H23N
UN Number UN3077
Health Hazard 1 Danger
Melting Point 148° to 153°C
Recommended Storage 2° to 8°C
Synonym (S)-2-(Boc-amino)-4-methyl-4-pentenoic acid dicyclohexylamine salt; Boc-4,5-dehydro-Leu-OH.DCHA
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C