Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-4,4-difluoro-L-proline methyl ester, 98%

$107.43 - $338.64

Chemical Identifiers

Chemical Name or Material N-Boc-4,4-difluoro-L-proline methyl ester
CAS 203866-17-5
Molecular Formula C11H17F2NO4
Formula Weight 265.26
MDL Number MFCD03094918
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2772703

  SDS
alfa aesar™
H2772703
1g Each for $338.64

AAH27727MD

  SDS
alfa aesar™
H27727MD
250mg Each for $107.43
Description & Specifications

Specifications

Formula Weight 265.26
Chemical Name or Material N-Boc-4,4-difluoro-L-proline methyl ester
Assay 98%
Molecular Formula C11H17F2NO4
MDL Number MFCD03094918
CAS 203866-17-5
Quantity 1g
Recommended Storage 2° to 8°C
Safety and Handling

2° to 8°C