Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-4,4-difluoro-L-proline, 97+%

$146.46 - $373.68

Chemical Identifiers

Chemical Name or Material N-Boc-4,4-difluoro-L-proline
CAS 203866-15-3
Molecular Formula C10H15F2NO4
Formula Weight 251.24
MDL Number MFCD03094917
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2705203

  SDS
alfa aesar™
H2705203
1g Each for $373.68

AAH27052MD

  SDS
alfa aesar™
H27052MD
250mg Each for $146.46
Description & Specifications

Specifications

Formula Weight 251.24
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material N-Boc-4,4-difluoro-L-proline
Molecular Formula C10H15F2NO4
Melting Point 120° to 124°C
MDL Number MFCD03094917
CAS 203866-15-3
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
Quantity 1g
Assay ≥97%
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.