Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-3-trifluoromethyl-L-phenylalanine, 98%

Chemical Identifiers

Chemical Name or Material N-Boc-3-trifluoromethyl-L-phenylalanine
CAS 142995-31-1
Formula Weight 333.3
MDL Number MFCD00672524
Beilstein 7048145
Synonym Boc-L-3-trifluoromethylphe; Boc-Phe(3-CF3)-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6263703

  SDS
alfa aesar™
H6263703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6263706

  SDS
alfa aesar™
H6263706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 333.3
Melting Point 135° to 138°C
Chemical Name or Material N-Boc-3-trifluoromethyl-L-phenylalanine
Physical Form Solid
Assay 98%
MDL Number MFCD00672524
CAS 142995-31-1
Beilstein 7048145
Synonym Boc-L-3-trifluoromethylphe; Boc-Phe(3-CF3)-OH
Quantity 1g