Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-3-pyrroline, 97%

$176.30 - $664.49

Chemical Identifiers

Chemical Name or Material N-Boc-3-pyrroline
CAS 73286-70-1
Molecular Formula C9H15NO2
Formula Weight 169.22
MDL Number MFCD01863512
Synonym N-Boc-2,5-dihydro-1H-pyrrole; tert-Butyl 2,5-dihydro-1H-pyrrole-1-carboxylate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2040606

  SDS
alfa aesar™
L2040606
5g Each for $664.49

AAL2040603

  SDS
alfa aesar™
L2040603
1g Each for $176.30
Description & Specifications

Specifications

Formula Weight 169.22
Chemical Name or Material N-Boc-3-pyrroline
Assay 97%
CAS 73286-70-1
Molecular Formula C9H15NO2
MDL Number MFCD01863512
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Boiling Point 208°C
Quantity 5g
Flash Point 81°C (178°F)
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Synonym N-Boc-2,5-dihydro-1H-pyrrole; tert-Butyl 2,5-dihydro-1H-pyrrole-1-carboxylate

Applications
Used in a synthesis of ß-aryl-GABA analogues by Heck arylation with arenediazonium salts, as well as synthesize aryl pyrrolizidines. N-Boc-pyrroline has also been utilized in the synthesis of various antibacterial compounds. It has also been used in the preparation of regioisomeric 3-hydroxyisoxazolinyl prolines which are medicinally active.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.