Please login to your online account to display your discounted pricing

N-BOC-3-Pyrroline, 96%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC427710050

 View more versions of this product

Catalog No. AC427710050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Low Melting Solid or Liquid
Assay ≥95%
Formula Weight 169.22
Quantity 5g
Color White to yellow
Density 0.981g/mL
CAS Min % 95.0
CAS Max % 100.0
Chemical Name or Material N-BOC-3-Pyrroline
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Assay Percent Range 96%
Molecular Formula C9H15NO2
MDL Number MFCD01863512
CAS 73286-70-1
NMR Consistent with structure
Synonym N-BOC-2, 5-Dihydro-1H-pyrrole
Infrared Spectrum Authentic

Specifications

Chemical Name or Material N-BOC-3-Pyrroline
CAS 73286-70-1
CAS Min % 95.0
CAS Max % 100.0
Assay ≥95%
Assay Percent Range 96%
Molecular Formula C9H15NO2
Formula Weight 169.22
MDL Number MFCD01863512
Synonym N-BOC-2, 5-Dihydro-1H-pyrrole
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents