missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-BOC-3-piperidone, 97%, ACROS Organics™

Chemical Identifiers

CAS 98977-36-7
Molecular Formula C10H17NO3
Molecular Weight (g/mol) 199.25
MDL Number MFCD01631193
InChI Key RIFXIGDBUBXKEI-UHFFFAOYSA-N
Synonym 1-boc-3-piperidone, 1-boc-3-piperidinone, n-boc-3-piperidone, 1-n-boc-3-piperidone, n-boc-3-piperidinone, boc-3-piperidone, 1-tert-butoxycarbonyl-3-piperidone, 3-oxo-piperidine-1-carboxylic acid tert-butyl ester, 3-oxopiperidine, n-boc protected, tert-butyl 3-oxopiperidinecarboxylate
PubChem CID 2756825
IUPAC Name tert-butyl 3-oxopiperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC(=O)C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC397001000 SDS View Documents Acros Organics
AC397001000
100g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC397000050 SDS View Documents Acros Organics
AC397000050
5g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC397000250 SDS View Documents Acros Organics
AC397000250
25g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

98977-36-7
199.25
RIFXIGDBUBXKEI-UHFFFAOYSA-N
2756825
CC(C)(C)OC(=O)N1CCCC(=O)C1
C10H17NO3
MFCD01631193
1-boc-3-piperidone, 1-boc-3-piperidinone, n-boc-3-piperidone, 1-n-boc-3-piperidone, n-boc-3-piperidinone, boc-3-piperidone, 1-tert-butoxycarbonyl-3-piperidone, 3-oxo-piperidine-1-carboxylic acid tert-butyl ester, 3-oxopiperidine, n-boc protected, tert-butyl 3-oxopiperidinecarboxylate
tert-butyl 3-oxopiperidine-1-carboxylate

Specifications

Authentic
Colorless
98977-36-7
100.0
C10H17NO3
1-boc-3-piperidone, 1-boc-3-piperidinone, n-boc-3-piperidone, 1-n-boc-3-piperidone, n-boc-3-piperidinone, boc-3-piperidone, 1-tert-butoxycarbonyl-3-piperidone, 3-oxo-piperidine-1-carboxylic acid tert-butyl ester, 3-oxopiperidine, n-boc protected, tert-butyl 3-oxopiperidinecarboxylate
CC(C)(C)OC(=O)N1CCCC(=O)C1
199.25
199.24
≥96.0% (GC)
Glass bottle
100g
96.0
97%
MFCD01631193
RIFXIGDBUBXKEI-UHFFFAOYSA-N
tert-butyl 3-oxopiperidine-1-carboxylate
2756825
Liquid
N-BOC-3-piperidone
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.