missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Boc-3-methyl-D-phenylalanine, 95%, Thermo Scientific™
$129.70 - $129.70
Chemical Identifiers
CAS | 114873-14-2 |
---|---|
Molecular Formula | C15H21NO4 |
Molecular Weight (g/mol) | 279.336 |
MDL Number | MFCD01862943 |
InChI Key | HBBXWMALJNZDOM-GFCCVEGCSA-N |
Synonym | boc-3-methyl-d-phenylalanine, boc-d-phe 3-me-oh, boc-d-3-methylphenylalanine, r-2-tert-butoxycarbonyl amino-3-m-tolyl propanoic acid, boc-d-3-methylphe, boc-3-methy-d-phenylalanine, 2r-2-tert-butoxycarbonyl amino-3-3-methylphenyl propanoic acid, boc-m-me-d-phe-oh, boc-d-3-me-phe-oh, boc-d-3-methyl phenylalanine |
PubChem CID | 7010034 |
IUPAC Name | (2R)-3-(3-methylphenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid |
SMILES | CC1=CC(=CC=C1)CC(C(=O)O)NC(=O)OC(C)(C)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6261203
|
Thermo Scientific Chemicals
H6261203 |
1 g |
N/A
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
114873-14-2 | |
279.336 | |
HBBXWMALJNZDOM-GFCCVEGCSA-N | |
7010034 | |
CC1=CC(=CC=C1)CC(C(=O)O)NC(=O)OC(C)(C)C |
C15H21NO4 | |
MFCD01862943 | |
boc-3-methyl-d-phenylalanine, boc-d-phe 3-me-oh, boc-d-3-methylphenylalanine, r-2-tert-butoxycarbonyl amino-3-m-tolyl propanoic acid, boc-d-3-methylphe, boc-3-methy-d-phenylalanine, 2r-2-tert-butoxycarbonyl amino-3-3-methylphenyl propanoic acid, boc-m-me-d-phe-oh, boc-d-3-me-phe-oh, boc-d-3-methyl phenylalanine | |
(2R)-3-(3-methylphenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid |
Specifications
114873-14-2 | |
MFCD01862943 | |
boc-3-methyl-d-phenylalanine, boc-d-phe 3-me-oh, boc-d-3-methylphenylalanine, r-2-tert-butoxycarbonyl amino-3-m-tolyl propanoic acid, boc-d-3-methylphe, boc-3-methy-d-phenylalanine, 2r-2-tert-butoxycarbonyl amino-3-3-methylphenyl propanoic acid, boc-m-me-d-phe-oh, boc-d-3-me-phe-oh, boc-d-3-methyl phenylalanine | |
CC1=CC(=CC=C1)CC(C(=O)O)NC(=O)OC(C)(C)C | |
279.336 | |
279.33 | |
N-Boc-3-methyl-D-phenylalanine |
C15H21NO4 | |
1 g | |
HBBXWMALJNZDOM-GFCCVEGCSA-N | |
(2R)-3-(3-methylphenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid | |
7010034 | |
95% |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures