missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-3-iodo-L-alanine methyl ester, 98%

Manufacturer:  Alfa Aesar™ H61638MD

 View more versions of this product

Catalog No. AAH61638MD



Description

Description

Applications
N-Boc-3-iodo-L-alanine methyl ester is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Soluble in water and 1% acetic acid.

Notes
Light sensitive. Store in dark. Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Specifications

Specifications

N-Boc-3-iodo-L-alanine methyl ester
C9H16INO4
4422011
UGZBFCCHLUWCQI-LURJTMIESA-N
methyl (2R)-3-iodo-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate
10903591
Powder
White
250mg
93267-04-0
MFCD00216579
boc-beta-iodo-ala-ome, methyl 2r-2-tert-butoxycarbonyl amino-3-iodopropanoate, boc-3-iodo-l-alanine methyl ester, r-methyl 2-tert-butoxycarbonyl amino-3-iodopropanoate, n-tert-butoxycarbonyl-3-iodo-l-alanine methyl ester, l-n-boc-3-iodoalanine methyl ester, r-methyl 2-tert-butoxycarbonylamino-3-iodopropanoate, boc-3-iodo-l-ala-ome, n-boc-3-iodo-l-alanine methyl ester, r-boc-, a-iodo-ala-ome
CC(C)(C)OC(=O)NC(CI)C(=O)OC
329.134
329.13
98%
50°C to 52°C
Light Sensitive

Chemical Identifiers

93267-04-0
329.134
UGZBFCCHLUWCQI-LURJTMIESA-N
10903591
CC(C)(C)OC(=O)NC(CI)C(=O)OC
C9H16INO4
MFCD00216579
boc-beta-iodo-ala-ome, methyl 2r-2-tert-butoxycarbonyl amino-3-iodopropanoate, boc-3-iodo-l-alanine methyl ester, r-methyl 2-tert-butoxycarbonyl amino-3-iodopropanoate, n-tert-butoxycarbonyl-3-iodo-l-alanine methyl ester, l-n-boc-3-iodoalanine methyl ester, r-methyl 2-tert-butoxycarbonylamino-3-iodopropanoate, boc-3-iodo-l-ala-ome, n-boc-3-iodo-l-alanine methyl ester, r-boc-, a-iodo-ala-ome
methyl (2R)-3-iodo-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C

Documents