missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-3-iodo-L-alanine methyl ester, 98%

Manufacturer:  Alfa Aesar™ H6163806

 View more versions of this product

Catalog No. AAH6163806



Description

Description

Applications
N-Boc-3-iodo-L-alanine methyl ester is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Soluble in water and 1% acetic acid.

Notes
Light sensitive. Store in dark. Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Specifications

Specifications

N-Boc-3-iodo-L-alanine methyl ester
C9H16INO4
4422011
Soluble in water and 1% acetic acid.
CC(C)(C)OC(=O)NC(CI)C(=O)OC
329.134
329.13
98%
50°C to 52°C
Light sensitive
93267-04-0
MFCD00216579
a-iodo-ala-ome
UGZBFCCHLUWCQI-LURJTMIESA-N
methyl (2R)-3-iodo-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate
10903591
Powder
White
5g

Chemical Identifiers

93267-04-0
329.134
UGZBFCCHLUWCQI-LURJTMIESA-N
10903591
CC(C)(C)OC(=O)NC(CI)C(=O)OC
C9H16INO4
MFCD00216579
a-iodo-ala-ome
methyl (2R)-3-iodo-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

Documents