missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-3-cyclohexyl-L-alanine, 98%

Manufacturer:  Alfa Aesar™ H5528203

 View more versions of this product

Catalog No. AAH5528203



Description

Description

Applications
It is an agrochemical, organic and pharmaceutical intermediates.

Notes
Storage: Keep container tightly closed. Store in a cool, dark and well-ventilated place and away from oxidizing agents.
Specifications

Specifications

N-Boc-3-cyclohexyl-L-alanine
37736-82-6
MFCD00211335
2s-2-tert-butoxycarbonyl amino-3-cyclohexylpropanoic acid
CC(C)(C)OC(=O)NC(CC1CCCCC1)C(=O)O
271.357
271.36
1g
C14H25NO4
3060603
MSZQAQJBXGTSHP-NSHDSACASA-N
(2S)-3-cyclohexyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
10588189
98%

Chemical Identifiers

37736-82-6
271.357
MSZQAQJBXGTSHP-NSHDSACASA-N
10588189
CC(C)(C)OC(=O)NC(CC1CCCCC1)C(=O)O
C14H25NO4
MFCD00211335
2s-2-tert-butoxycarbonyl amino-3-cyclohexylpropanoic acid
(2S)-3-cyclohexyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

Documents