missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-3-cyclohexyl-D-alanine, 98%

Manufacturer:  Alfa Aesar™ H55339MD

 View more versions of this product

Catalog No. AAH55339MD



Specifications

Specifications

N-Boc-3-cyclohexyl-D-alanine
250mg
C14H25NO4
4192671
MSZQAQJBXGTSHP-LLVKDONJSA-N
(2R)-3-cyclohexyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
2755927
98%
64°C to 67°C
127095-92-5
MFCD00211334
2r-2-tert-butoxy carbonyl amino-3-cyclohexylpropanoic acid
CC(C)(C)OC(=O)NC(CC1CCCCC1)C(=O)O
271.357
271.36

Chemical Identifiers

127095-92-5
271.357
MSZQAQJBXGTSHP-LLVKDONJSA-N
2755927
CC(C)(C)OC(=O)NC(CC1CCCCC1)C(=O)O
C14H25NO4
MFCD00211334
2r-2-tert-butoxy carbonyl amino-3-cyclohexylpropanoic acid
(2R)-3-cyclohexyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

Documents