missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-3-cyano-D-phenylalanine, 95%

$44.86 - $590.64

Chemical Identifiers

CAS 205445-56-3
Molecular Formula C15H18N2O4
Molecular Weight (g/mol) 290.319
MDL Number MFCD00797561
InChI Key FDQDHMZKOPOWFE-GFCCVEGCSA-N
Synonym boc-d-phe 3-cn-oh, boc-d-3-cyanophenylalanine, boc-3-cyano-d-phenylalanine, r-2-tert-butoxycarbonyl amino-3-3-cyanophenyl propanoic acid, 2r-2-tert-butoxycarbonyl amino-3-3-cyanophenyl propanoic acid, boc-d-3-cyanophe, boc-d-3-cn-phe-oh, boc-d-3-cyano phenylalanine, ksc913m8f, r-n-boc-3-3cyanophenylalanine
PubChem CID 7021063
IUPAC Name (2R)-3-(3-cyanophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NC(CC1=CC=CC(=C1)C#N)C(=O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH62038MD View Documents Alfa Aesar™
H62038MD
250mg Each for $44.86
 
AAH6203803 View Documents Alfa Aesar™
H6203803
1g Each for $132.77
 
AAH6203806 View Documents Alfa Aesar™
H6203806
5g Each for $590.64
 
Specifications

Chemical Identifiers

205445-56-3
290.319
FDQDHMZKOPOWFE-GFCCVEGCSA-N
7021063
CC(C)(C)OC(=O)NC(CC1=CC=CC(=C1)C#N)C(=O)O
C15H18N2O4
MFCD00797561
boc-d-phe 3-cn-oh, boc-d-3-cyanophenylalanine, boc-3-cyano-d-phenylalanine, r-2-tert-butoxycarbonyl amino-3-3-cyanophenyl propanoic acid, 2r-2-tert-butoxycarbonyl amino-3-3-cyanophenyl propanoic acid, boc-d-3-cyanophe, boc-d-3-cn-phe-oh, boc-d-3-cyano phenylalanine, ksc913m8f, r-n-boc-3-3cyanophenylalanine
(2R)-3-(3-cyanophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid

Specifications

250mg
C15H18N2O4
UN3439
FDQDHMZKOPOWFE-GFCCVEGCSA-N
(2R)-3-(3-cyanophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
7021063
95%
205445-56-3
MFCD00797561
boc-d-phe 3-cn-oh, boc-d-3-cyanophenylalanine, boc-3-cyano-d-phenylalanine, r-2-tert-butoxycarbonyl amino-3-3-cyanophenyl propanoic acid, 2r-2-tert-butoxycarbonyl amino-3-3-cyanophenyl propanoic acid, boc-d-3-cyanophe, boc-d-3-cn-phe-oh, boc-d-3-cyano phenylalanine, ksc913m8f, r-n-boc-3-3cyanophenylalanine
CC(C)(C)OC(=O)NC(CC1=CC=CC(=C1)C#N)C(=O)O
290.319
290.31
N-Boc-3-cyano-D-phenylalanine
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
P261-P280-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.