Learn More
N-Boc-3-cyano-D-phenylalanine, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H62038MD
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
N-Boc-3-cyano-D-phenylalanine | |
C15H18N2O4 | |
250 mg | |
boc-d-phe 3-cn-oh, boc-d-3-cyanophenylalanine, boc-3-cyano-d-phenylalanine, r-2-tert-butoxycarbonyl amino-3-3-cyanophenyl propanoic acid, 2r-2-tert-butoxycarbonyl amino-3-3-cyanophenyl propanoic acid, boc-d-3-cyanophe, boc-d-3-cn-phe-oh, boc-d-3-cyano phenylalanine, ksc913m8f, r-n-boc-3-3cyanophenylalanine | |
CC(C)(C)OC(=O)NC(CC1=CC=CC(=C1)C#N)C(=O)O | |
290.319 | |
290.31 |
205445-56-3 | |
MFCD00797561 | |
UN3439 | |
FDQDHMZKOPOWFE-GFCCVEGCSA-N | |
(2R)-3-(3-cyanophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid | |
7021063 | |
95% |
Chemical Identifiers
205445-56-3 | |
290.319 | |
FDQDHMZKOPOWFE-GFCCVEGCSA-N | |
7021063 | |
CC(C)(C)OC(=O)NC(CC1=CC=CC(=C1)C#N)C(=O)O |
C15H18N2O4 | |
MFCD00797561 | |
boc-d-phe 3-cn-oh, boc-d-3-cyanophenylalanine, boc-3-cyano-d-phenylalanine, r-2-tert-butoxycarbonyl amino-3-3-cyanophenyl propanoic acid, 2r-2-tert-butoxycarbonyl amino-3-3-cyanophenyl propanoic acid, boc-d-3-cyanophe, boc-d-3-cn-phe-oh, boc-d-3-cyano phenylalanine, ksc913m8f, r-n-boc-3-3cyanophenylalanine | |
(2R)-3-(3-cyanophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid |
Safety and Handling
GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
H302+H312+H332
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures