Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-3-bromo-D-phenylalanine, 98%

Chemical Identifiers

Chemical Name or Material N-Boc-3-bromo-D-phenylalanine
CAS 261360-77-4
Formula Weight 344.2
MDL Number MFCD01317704
Synonym (R)-N-Boc-3-bromophenylalanine; Boc-Phe(3-Br)-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6276103

 
alfa aesar™
H6276103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6276106

 
alfa aesar™
H6276106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH62761MD

 
alfa aesar™
H62761MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 344.2
Chemical Name or Material N-Boc-3-bromo-D-phenylalanine
Physical Form Powder
Assay 98%
MDL Number MFCD01317704
CAS 261360-77-4
Synonym (R)-N-Boc-3-bromophenylalanine; Boc-Phe(3-Br)-OH
Color White
Quantity 1g
Storage Note 1 2° to 8°C
Safety and Handling

2° to 8°C