missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-3-bromo-D-phenylalanine, 98%

Manufacturer:  Alfa Aesar™ H6276106

 View more versions of this product

Catalog No. AAH6276106


 

Specifications

Specifications

N-Boc-3-bromo-D-phenylalanine
98%
MFCD01317704
FBUDYESOPLBQIR-LLVKDONJSA-N
(2R)-3-(3-bromophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
7006629
Powder
5g
261360-77-4
C14H18BrNO4
(R)-N-Boc-3-bromophenylalanine; Boc-Phe(3-Br)-OH
CC(C)(C)OC(=O)NC(CC1=CC(=CC=C1)Br)C(=O)O
344.205
344.2
White

Chemical Identifiers

261360-77-4
344.205
FBUDYESOPLBQIR-LLVKDONJSA-N
7006629
C14H18BrNO4
MFCD01317704
(R)-N-Boc-3-bromophenylalanine; Boc-Phe(3-Br)-OH
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C

SDS Documents