Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-3-bromo-2-isoxazoline-5-methylamine, 97%

$195.49 - $597.24

Chemical Identifiers

Chemical Name or Material N-Boc-3-bromo-2-isoxazoline-5-methylamine
CAS 109770-82-3
Molecular Formula C9H15BrN2O3
Formula Weight 279.14
MDL Number MFCD08741480
Synonym (3-Bromo-4,5-dihydro-5-isoxazolylmethyl)carbamic acid tert-butyl ester; N-tert-Butoxycarbonyl-3-bromo-2-isoxazolidine-5-methylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2627427

 
alfa aesar™
H2627427
2.5g Each for $597.24

AAH26274ME

 
alfa aesar™
H26274ME
500mg Each for $195.49
Description & Specifications

Specifications

Formula Weight 279.14
Chemical Name or Material N-Boc-3-bromo-2-isoxazoline-5-methylamine
Assay 97%
Molecular Formula C9H15BrN2O3
MDL Number MFCD08741480
CAS 109770-82-3
Quantity 2.5g
Synonym (3-Bromo-4,5-dihydro-5-isoxazolylmethyl)carbamic acid tert-butyl ester; N-tert-Butoxycarbonyl-3-bromo-2-isoxazolidine-5-methylamine