missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-3-benzyloxymethyl-L-histidine, 98%

$32.62 - $482.16

Chemical Identifiers

CAS 79950-65-5
MDL Number MFCD00043133
Synonym Boc-His(Bom)-OH; N-[(1,1-Dimethylethoxy)carbonyl]-3-[(phenylmethoxy)methyl]- L-histidine
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6123403
View Documents
Alfa Aesar™
H6123403
1g Each for $32.62
 
AAH6123406
View Documents
Alfa Aesar™
H6123406
5g Each for $121.19
 
AAH6123414
View Documents
Alfa Aesar™
H6123414
25g Each for $482.16
 
Specifications

Chemical Identifiers

79950-65-5
MFCD00043133
Boc-His(Bom)-OH; N-[(1,1-Dimethylethoxy)carbonyl]-3-[(phenylmethoxy)methyl]- L-histidine

Specifications

79950-65-5
4335455
375.42
98%
1g
MFCD00043133
Boc-His(Bom)-OH; N-[(1,1-Dimethylethoxy)carbonyl]-3-[(phenylmethoxy)methyl]- L-histidine
Powder
White
N-Boc-3-benzyloxymethyl-L-histidine
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Keep cold