missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Boc-3-(4-pyridyl)-L-alanine, 95%, Thermo Scientific™
$98.26 - $98.26
Chemical Identifiers
CAS | 37535-57-2 |
---|---|
Molecular Formula | C13H18N2O4 |
Molecular Weight (g/mol) | 266.297 |
MDL Number | MFCD00672528 |
InChI Key | FNYWDMKESUACOU-JTQLQIEISA-N |
Synonym | boc-l-4-pyridylalanine, boc-ala 4-pyridyl-oh, boc-4-pal-oh, s-2-tert-butoxycarbonyl amino-3-pyridin-4-yl propanoic acid, boc-3-4-pyridyl-l-alanine, boc-l-3-4-pyridyl-alanine, boc-l-4-pyridylala, boc-3-4-pyridyl-ala-oh, boc-4'-pyridyl-l-ala, s-n-boc-4-pyridyl alanine |
PubChem CID | 7020844 |
IUPAC Name | (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-pyridin-4-ylpropanoic acid |
SMILES | CC(C)(C)OC(=O)NC(CC1=CC=NC=C1)C(=O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH62532MD
|
Thermo Scientific Chemicals
H62532MD |
250 mg |
Each for $98.26
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
37535-57-2 | |
266.297 | |
FNYWDMKESUACOU-JTQLQIEISA-N | |
7020844 | |
CC(C)(C)OC(=O)NC(CC1=CC=NC=C1)C(=O)O |
C13H18N2O4 | |
MFCD00672528 | |
boc-l-4-pyridylalanine, boc-ala 4-pyridyl-oh, boc-4-pal-oh, s-2-tert-butoxycarbonyl amino-3-pyridin-4-yl propanoic acid, boc-3-4-pyridyl-l-alanine, boc-l-3-4-pyridyl-alanine, boc-l-4-pyridylala, boc-3-4-pyridyl-ala-oh, boc-4'-pyridyl-l-ala, s-n-boc-4-pyridyl alanine | |
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-pyridin-4-ylpropanoic acid |
Specifications
37535-57-2 | |
MFCD00672528 | |
boc-l-4-pyridylalanine, boc-ala 4-pyridyl-oh, boc-4-pal-oh, s-2-tert-butoxycarbonyl amino-3-pyridin-4-yl propanoic acid, boc-3-4-pyridyl-l-alanine, boc-l-3-4-pyridyl-alanine, boc-l-4-pyridylala, boc-3-4-pyridyl-ala-oh, boc-4'-pyridyl-l-ala, s-n-boc-4-pyridyl alanine | |
CC(C)(C)OC(=O)NC(CC1=CC=NC=C1)C(=O)O | |
266.297 | |
266.29 | |
250 mg | |
N-Boc-3-(4-pyridyl)-L-alanine |
C13H18N2O4 | |
6417084 | |
FNYWDMKESUACOU-JTQLQIEISA-N | |
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-pyridin-4-ylpropanoic acid | |
7020844 | |
95% | |
Air and moisture sensitive |
Safety and Handling
TSCA : No
Recommended Storage : Store at -20°C; Store under Argon