missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-BOC-3-(4-Cyanophenyl)oxaziridine, 98+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 297095000

 View more versions of this product

Catalog No. AC297095000


 

Specifications Description & Specifications

Specifications

246.27
500mg
57° to 63°C
100.0
98+%
C13H14N2O3
150884-56-3
Glass bottle
Crystalline Powder
White to very slightly beige
98.0
N-BOC-3-(4-Cyanophenyl)oxaziridine
MFCD00800493
98+%
Authentic
98.0% min.

Chemical Identifiers

150884-56-3
100.0
C13H14N2O3
MFCD00800493
98.0
98+%
246.27
N-BOC-3-(4-Cyanophenyl)oxaziridine, 98+%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning