missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-3-(4-biphenylyl)-D-alanine, 95%

Manufacturer:  Alfa Aesar™ H6349403

 View more versions of this product

Catalog No. AAH6349403



Specifications

Specifications

N-Boc-3-(4-biphenylyl)-D-alanine
C20H23NO4
boc-d-4,4'-biphenylalanine, boc-d-4,4-biphenylalanine, boc-3-4-biphenylyl-d-alanine, r-3-1,1'-biphenyl-4-yl-2-tert-butoxycarbonyl amino propanoic acid, boc-d-bip-oh, boc-d-4-phenylphenylalanine, boc-4-biphenyl-d-ala, boc-4-phenyl-d-phe-oh, boc-d-bph-oh, boc-d-bip 4,4'-oh
CC(C)(C)OC(=O)NC(CC1=CC=C(C=C1)C2=CC=CC=C2)C(=O)O
341.407
341.41
95%
128779-47-5
MFCD00191185
NBVVKAUSAGHTSU-QGZVFWFLSA-N
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-(4-phenylphenyl)propanoic acid
7019133
Solid
1g

Chemical Identifiers

128779-47-5
341.407
NBVVKAUSAGHTSU-QGZVFWFLSA-N
7019133
CC(C)(C)OC(=O)NC(CC1=CC=C(C=C1)C2=CC=CC=C2)C(=O)O
C20H23NO4
MFCD00191185
boc-d-4,4'-biphenylalanine, boc-d-4,4-biphenylalanine, boc-3-4-biphenylyl-d-alanine, r-3-1,1'-biphenyl-4-yl-2-tert-butoxycarbonyl amino propanoic acid, boc-d-bip-oh, boc-d-4-phenylphenylalanine, boc-4-biphenyl-d-ala, boc-4-phenyl-d-phe-oh, boc-d-bph-oh, boc-d-bip 4,4'-oh
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-(4-phenylphenyl)propanoic acid
Safety and Handling

Safety and Handling

storageNote1 : -30° to -10°C

SDS Documents