missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-3-(3-methoxy-3-oxo-1-propen-1-yl)indole, 97%

$72.03 - $951.49

Chemical Identifiers

CAS 388631-93-4
Molecular Formula C17H19NO4
Molecular Weight (g/mol) 301.342
MDL Number MFCD03844705
InChI Key VSMVLFKTUMKNRW-MDZDMXLPSA-N
Synonym tert-butyl 3-3-methoxy-3-oxopropen-1-yl indole-1-carboxylate, 3-2-methoxycarbonyl-vinyl-indole-1-carboxylic acid tert-butyl ester, tert-butyl 3-1e-3-methoxy-3-oxoprop-1-en-1-yl indole-1-carboxylate, tert-butyl 3-3-methoxy-3-oxoprop-1-en-1-yl-1h-indole-1-carboxylate, 1-tert-butoxycarbonyl-1h-indole-3-acrylic acid methyl ester
PubChem CID 12190555
IUPAC Name tert-butyl 3-[(E)-3-methoxy-3-oxoprop-1-enyl]indole-1-carboxylate
SMILES CC(C)(C)OC(=O)N1C=C(C2=CC=CC=C21)C=CC(=O)OC
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH63712MD View Documents Alfa Aesar™
H63712MD
250mg Each for $72.03
 
AAH6371203 View Documents Alfa Aesar™
H6371203
1g Each for $229.77
 
AAH6371206 View Documents Alfa Aesar™
H6371206
5g Each for $951.49
 
Specifications

Chemical Identifiers

388631-93-4
301.342
VSMVLFKTUMKNRW-MDZDMXLPSA-N
12190555
CC(C)(C)OC(=O)N1C=C(C2=CC=CC=C21)C=CC(=O)OC
C17H19NO4
MFCD03844705
tert-butyl 3-3-methoxy-3-oxopropen-1-yl indole-1-carboxylate, 3-2-methoxycarbonyl-vinyl-indole-1-carboxylic acid tert-butyl ester, tert-butyl 3-1e-3-methoxy-3-oxoprop-1-en-1-yl indole-1-carboxylate, tert-butyl 3-3-methoxy-3-oxoprop-1-en-1-yl-1h-indole-1-carboxylate, 1-tert-butoxycarbonyl-1h-indole-3-acrylic acid methyl ester
tert-butyl 3-[(E)-3-methoxy-3-oxoprop-1-enyl]indole-1-carboxylate

Specifications

250mg
C17H19NO4
tert-butyl 3-3-methoxy-3-oxopropen-1-yl indole-1-carboxylate, 3-2-methoxycarbonyl-vinyl-indole-1-carboxylic acid tert-butyl ester, tert-butyl 3-1e-3-methoxy-3-oxoprop-1-en-1-yl indole-1-carboxylate, tert-butyl 3-3-methoxy-3-oxoprop-1-en-1-yl-1h-indole-1-carboxylate, 1-tert-butoxycarbonyl-1h-indole-3-acrylic acid methyl ester
CC(C)(C)OC(=O)N1C=C(C2=CC=CC=C21)C=CC(=O)OC
301.342
301.34
97%
388631-93-4
MFCD03844705
VSMVLFKTUMKNRW-MDZDMXLPSA-N
tert-butyl 3-[(E)-3-methoxy-3-oxoprop-1-enyl]indole-1-carboxylate
12190555
Solid
N-Boc-3-(3-methoxy-3-oxo-1-propen-1-yl)indole
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C