Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-3-(2-furyl)-L-alanine dicyclohexylammonium salt, 98%

Chemical Identifiers

Chemical Name or Material N-Boc-3-(2-furyl)-L-alanine dicyclohexylammonium salt
CAS 331730-08-6
Formula Weight 436.58
MDL Number MFCD01860894
Synonym Boc-β-(2-furyl)-Ala-OH.DCHA
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6225803

  SDS
alfa aesar™
H6225803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6225806

  SDS
alfa aesar™
H6225806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH62258MD

  SDS
alfa aesar™
H62258MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 436.58
Chemical Name or Material N-Boc-3-(2-furyl)-L-alanine dicyclohexylammonium salt
Physical Form Powder
Assay 98%
MDL Number MFCD01860894
CAS 331730-08-6
Synonym Boc-β-(2-furyl)-Ala-OH.DCHA
Color White
Quantity 1g
Storage Note 1 2° to 8°C
Safety and Handling

2° to 8°C