missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-2-trifluoromethyl-L-phenylalanine, 95%

$76.65 - $245.20

Chemical Identifiers

CAS 167993-21-7
Molecular Formula C15H18F3NO4
Molecular Weight (g/mol) 333.307
MDL Number MFCD01862946
InChI Key XKMOOODKNPYTEE-NSHDSACASA-N
Synonym boc-2-trifluoromethyl-l-phenylalanine, boc-l-2-trifluoromethylphenylalanine, boc-2-trifluoromethyl-l-phenylalanine, s-2-tert-butoxycarbonyl amino-3-2-trifluoromethyl phenyl propanoic acid, boc-phe 2-cf3-oh, boc-l-2-trifluoromethylphe, 2s-2-tert-butoxycarbonyl amino-3-2-trifluoromethyl phenyl propanoic acid, boc-phe 2-cf 3-oh, 2s-2-tert-butoxycarbonylamino-3-2-trifluoromethyl phenyl propanoic acid, 2-trifluoromethyl-l-phenylalanine, n-boc protected
PubChem CID 2761473
IUPAC Name (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-[2-(trifluoromethyl)phenyl]propanoic acid
SMILES CC(C)(C)OC(=O)NC(CC1=CC=CC=C1C(F)(F)F)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH62836MD View Documents Alfa Aesar™
H62836MD
250mg Each for $76.65
 
AAH6283603 View Documents Alfa Aesar™
H6283603
1g Each for $245.20
 
Specifications

Chemical Identifiers

167993-21-7
333.307
XKMOOODKNPYTEE-NSHDSACASA-N
2761473
CC(C)(C)OC(=O)NC(CC1=CC=CC=C1C(F)(F)F)C(=O)O
C15H18F3NO4
MFCD01862946
boc-2-trifluoromethyl-l-phenylalanine, boc-l-2-trifluoromethylphenylalanine, boc-2-trifluoromethyl-l-phenylalanine, s-2-tert-butoxycarbonyl amino-3-2-trifluoromethyl phenyl propanoic acid, boc-phe 2-cf3-oh, boc-l-2-trifluoromethylphe, 2s-2-tert-butoxycarbonyl amino-3-2-trifluoromethyl phenyl propanoic acid, boc-phe 2-cf 3-oh, 2s-2-tert-butoxycarbonylamino-3-2-trifluoromethyl phenyl propanoic acid, 2-trifluoromethyl-l-phenylalanine, n-boc protected
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-[2-(trifluoromethyl)phenyl]propanoic acid

Specifications

250mg
C15H18F3NO4
boc-2-trifluoromethyl-l-phenylalanine, boc-l-2-trifluoromethylphenylalanine, boc-2-trifluoromethyl-l-phenylalanine, s-2-tert-butoxycarbonyl amino-3-2-trifluoromethyl phenyl propanoic acid, boc-phe 2-cf3-oh, boc-l-2-trifluoromethylphe, 2s-2-tert-butoxycarbonyl amino-3-2-trifluoromethyl phenyl propanoic acid, boc-phe 2-cf 3-oh, 2s-2-tert-butoxycarbonylamino-3-2-trifluoromethyl phenyl propanoic acid, 2-trifluoromethyl-l-phenylalanine, n-boc protected
CC(C)(C)OC(=O)NC(CC1=CC=CC=C1C(F)(F)F)C(=O)O
333.307
333.3
N-Boc-2-trifluoromethyl-L-phenylalanine
167993-21-7
MFCD01862946
XKMOOODKNPYTEE-NSHDSACASA-N
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-[2-(trifluoromethyl)phenyl]propanoic acid
2761473
95%
Safety and Handling

Safety and Handling

Recommended Storage : −30° to −10°C