missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-2-trifluoromethyl-D-phenylalanine, 95%

Manufacturer:  Alfa Aesar™ H6234803

 View more versions of this product

Catalog No. AAH6234803



Specifications

Specifications

N-Boc-2-trifluoromethyl-D-phenylalanine
C15H18F3NO4
2r-2-tert-butoxycarbonyl amino-3-2-trifluoromethyl phenyl propanoic acid
CC(C)(C)OC(=O)NC(CC1=CC=CC=C1C(F)(F)F)C(=O)O
333.307
333.3
95%
346694-78-8
MFCD01862945
XKMOOODKNPYTEE-LLVKDONJSA-N
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-[2-(trifluoromethyl)phenyl]propanoic acid
7010037
Solid
1g

Chemical Identifiers

346694-78-8
333.307
XKMOOODKNPYTEE-LLVKDONJSA-N
7010037
CC(C)(C)OC(=O)NC(CC1=CC=CC=C1C(F)(F)F)C(=O)O
C15H18F3NO4
MFCD01862945
2r-2-tert-butoxycarbonyl amino-3-2-trifluoromethyl phenyl propanoic acid
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-[2-(trifluoromethyl)phenyl]propanoic acid
Documents