Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-2-trifluoromethyl-D-phenylalanine, 95%

Chemical Identifiers

Chemical Name or Material N-Boc-2-trifluoromethyl-D-phenylalanine
CAS 346694-78-8
Formula Weight 333.3
MDL Number MFCD01862945
Synonym Boc-D-Phe(2-CF3)-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6234803

 
alfa aesar™
H6234803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6234806

 
alfa aesar™
H6234806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH62348MD

 
alfa aesar™
H62348MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 333.3
Physical Form Solid
CAS 346694-78-8
Quantity 1g
Chemical Name or Material N-Boc-2-trifluoromethyl-D-phenylalanine
Synonym Boc-D-Phe(2-CF3)-OH
MDL Number MFCD01862945
Assay 95%