missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-2-propargyl-D-glycine, 95%

$229.77 - $686.24

Chemical Identifiers

CAS 63039-46-3
Molecular Formula C10H15NO4
Molecular Weight (g/mol) 213.233
MDL Number MFCD01320886
InChI Key AMKHAJIFPHJYMH-SSDOTTSWSA-N
Synonym boc-d-propargylglycine, r-2-tert-butoxycarbonyl amino pent-4-ynoic acid, n-boc-2-propargyl-d-glycine, boc-d-pra-oh, s-n-boc-propargylglycine, 2r-2-tert-butoxycarbonyl amino pent-4-ynoic acid, boc-d-gly propargyl-oh, boc-r-2-propargylglycine, n-tert-butoxycarbonyl-3-ethynyl-d-alanine, 2-r-tert-butoxycarbonylaminopent-4-ynoic acid
PubChem CID 7009126
IUPAC Name (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pent-4-ynoic acid
SMILES CC(C)(C)OC(=O)NC(CC#C)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH62572MD View Documents Alfa Aesar™
H62572MD
250mg Each for $229.77
 
AAH6257203 View Documents Alfa Aesar™
H6257203
1g Each for $686.24
 
Specifications

Chemical Identifiers

63039-46-3
213.233
AMKHAJIFPHJYMH-SSDOTTSWSA-N
7009126
CC(C)(C)OC(=O)NC(CC#C)C(=O)O
C10H15NO4
MFCD01320886
boc-d-propargylglycine, r-2-tert-butoxycarbonyl amino pent-4-ynoic acid, n-boc-2-propargyl-d-glycine, boc-d-pra-oh, s-n-boc-propargylglycine, 2r-2-tert-butoxycarbonyl amino pent-4-ynoic acid, boc-d-gly propargyl-oh, boc-r-2-propargylglycine, n-tert-butoxycarbonyl-3-ethynyl-d-alanine, 2-r-tert-butoxycarbonylaminopent-4-ynoic acid
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pent-4-ynoic acid

Specifications

250mg
C10H15NO4
boc-d-propargylglycine, r-2-tert-butoxycarbonyl amino pent-4-ynoic acid, n-boc-2-propargyl-d-glycine, boc-d-pra-oh, s-n-boc-propargylglycine, 2r-2-tert-butoxycarbonyl amino pent-4-ynoic acid, boc-d-gly propargyl-oh, boc-r-2-propargylglycine, n-tert-butoxycarbonyl-3-ethynyl-d-alanine, 2-r-tert-butoxycarbonylaminopent-4-ynoic acid
CC(C)(C)OC(=O)NC(CC#C)C(=O)O
213.233
213.23
N-Boc-2-propargyl-D-glycine
63039-46-3
MFCD01320886
AMKHAJIFPHJYMH-SSDOTTSWSA-N
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pent-4-ynoic acid
7009126
95%