missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Boc-2-fluoro-L-phenylalanine, 98%, Thermo Scientific Chemicals
$124.59 - $455.08
Chemical Identifiers
CAS | 114873-00-6 |
---|---|
Molecular Formula | C14H18FNO4 |
Molecular Weight (g/mol) | 283.299 |
MDL Number | MFCD00672520 |
InChI Key | NTWUXBKUDXGMHV-NSHDSACASA-N |
Synonym | boc-phe 2-f-oh, boc-l-2-fluorophenylalanine, boc-2-fluoro-l-phenylalanine, boc-l-2-fluorophe, boc-l-phe 2-f-oh, s-n-boc-2-fluorophenylalanine, boc-d-2-fluorophenylalanine, s-2-tert-butoxycarbonyl amino-3-2-fluorophenyl propanoic acid, boc-2-fluoro-l-phe, 2s-2-tert-butoxycarbonyl amino-3-2-fluorophenyl propanoic acid |
PubChem CID | 2734496 |
IUPAC Name | (2S)-3-(2-fluorophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid |
SMILES | CC(C)(C)OC(=O)NC(CC1=CC=CC=C1F)C(=O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6262003
|
Thermo Scientific Chemicals
H6262003 |
1 g |
Each for $124.59
|
|
|||||
AAH6262006
|
Thermo Scientific Chemicals
H6262006 |
5 g |
Each for $455.08
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
114873-00-6 | |
283.299 | |
NTWUXBKUDXGMHV-NSHDSACASA-N | |
2734496 | |
CC(C)(C)OC(=O)NC(CC1=CC=CC=C1F)C(=O)O |
C14H18FNO4 | |
MFCD00672520 | |
boc-phe 2-f-oh, boc-l-2-fluorophenylalanine, boc-2-fluoro-l-phenylalanine, boc-l-2-fluorophe, boc-l-phe 2-f-oh, s-n-boc-2-fluorophenylalanine, boc-d-2-fluorophenylalanine, s-2-tert-butoxycarbonyl amino-3-2-fluorophenyl propanoic acid, boc-2-fluoro-l-phe, 2s-2-tert-butoxycarbonyl amino-3-2-fluorophenyl propanoic acid | |
(2S)-3-(2-fluorophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid |
Specifications
114873-00-6 | |
C14H18FNO4 | |
1 g | |
boc-phe 2-f-oh, boc-l-2-fluorophenylalanine, boc-2-fluoro-l-phenylalanine, boc-l-2-fluorophe, boc-l-phe 2-f-oh, s-n-boc-2-fluorophenylalanine, boc-d-2-fluorophenylalanine, s-2-tert-butoxycarbonyl amino-3-2-fluorophenyl propanoic acid, boc-2-fluoro-l-phe, 2s-2-tert-butoxycarbonyl amino-3-2-fluorophenyl propanoic acid | |
+9° (c=1 in Ethyl Acetate) | |
(2S)-3-(2-fluorophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid | |
2734496 | |
98% |
92°C to 96°C | |
MFCD00672520 | |
5347990 | |
NTWUXBKUDXGMHV-NSHDSACASA-N | |
CC(C)(C)OC(=O)NC(CC1=CC=CC=C1F)C(=O)O | |
283.299 | |
283.3 | |
N-Boc-2-fluoro-L-phenylalanine |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only