missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Boc-2-fluoro-L-phenylalanine, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6262006
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
N-Boc-2-fluoro-L-phenylalanine | |
92°C to 96°C | |
MFCD00672520 | |
5347990 | |
NTWUXBKUDXGMHV-NSHDSACASA-N | |
CC(C)(C)OC(=O)NC(CC1=CC=CC=C1F)C(=O)O | |
283.299 | |
283.3 |
114873-00-6 | |
C14H18FNO4 | |
5 g | |
boc-phe 2-f-oh, boc-l-2-fluorophenylalanine, boc-2-fluoro-l-phenylalanine, boc-l-2-fluorophe, boc-l-phe 2-f-oh, s-n-boc-2-fluorophenylalanine, boc-d-2-fluorophenylalanine, s-2-tert-butoxycarbonyl amino-3-2-fluorophenyl propanoic acid, boc-2-fluoro-l-phe, 2s-2-tert-butoxycarbonyl amino-3-2-fluorophenyl propanoic acid | |
+9° (c=1 in Ethyl Acetate) | |
(2S)-3-(2-fluorophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid | |
2734496 | |
98% |
Chemical Identifiers
114873-00-6 | |
283.299 | |
NTWUXBKUDXGMHV-NSHDSACASA-N | |
2734496 | |
CC(C)(C)OC(=O)NC(CC1=CC=CC=C1F)C(=O)O |
C14H18FNO4 | |
MFCD00672520 | |
boc-phe 2-f-oh, boc-l-2-fluorophenylalanine, boc-2-fluoro-l-phenylalanine, boc-l-2-fluorophe, boc-l-phe 2-f-oh, s-n-boc-2-fluorophenylalanine, boc-d-2-fluorophenylalanine, s-2-tert-butoxycarbonyl amino-3-2-fluorophenyl propanoic acid, boc-2-fluoro-l-phe, 2s-2-tert-butoxycarbonyl amino-3-2-fluorophenyl propanoic acid | |
(2S)-3-(2-fluorophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only