missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Boc-2-cyclopentyl-L-glycine dicyclohexylammonium salt, 95%, Thermo Scientific™
$152.48 - $152.48
Chemical Identifiers
CAS | 676559-50-5 |
---|---|
MDL Number | MFCD05663437 |
Synonym | boc-l-cyclopentylglycine, s-2-tert-butoxycarbonyl amino-2-cyclopentylacetic acid, s-tert-butoxycarbonylamino-cyclopentyl-acetic acid, s-tert-butoxycarbonyl amino cyclopentyl acetic acid, 2s-tert-butoxycarbonyl amino cyclopentyl ethanoic acid, 2s-2-tert-butoxy carbonyl amino-2-cyclopentylacetic acid, ambotzbaa5140, boc-l-cyclopentyl glycine, n-tert-butoxycarbonyl-l-cyclopentylglycine, n-alpha-t-butyloxycarbonyl-l-cyclopentylglycine |
PubChem CID | 7020746 |
IUPAC Name | (2S)-2-cyclopentyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]acetic acid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6283803
|
Thermo Scientific Chemicals
H6283803 |
1 g |
N/A
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
676559-50-5 | |
boc-l-cyclopentylglycine, s-2-tert-butoxycarbonyl amino-2-cyclopentylacetic acid, s-tert-butoxycarbonylamino-cyclopentyl-acetic acid, s-tert-butoxycarbonyl amino cyclopentyl acetic acid, 2s-tert-butoxycarbonyl amino cyclopentyl ethanoic acid, 2s-2-tert-butoxy carbonyl amino-2-cyclopentylacetic acid, ambotzbaa5140, boc-l-cyclopentyl glycine, n-tert-butoxycarbonyl-l-cyclopentylglycine, n-alpha-t-butyloxycarbonyl-l-cyclopentylglycine | |
(2S)-2-cyclopentyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]acetic acid |
MFCD05663437 | |
7020746 |
Specifications
676559-50-5 | |
C12H21NO4 | |
boc-l-cyclopentylglycine, s-2-tert-butoxycarbonyl amino-2-cyclopentylacetic acid, s-tert-butoxycarbonylamino-cyclopentyl-acetic acid, s-tert-butoxycarbonyl amino cyclopentyl acetic acid, 2s-tert-butoxycarbonyl amino cyclopentyl ethanoic acid, 2s-2-tert-butoxy carbonyl amino-2-cyclopentylacetic acid, ambotzbaa5140, boc-l-cyclopentyl glycine, n-tert-butoxycarbonyl-l-cyclopentylglycine, n-alpha-t-butyloxycarbonyl-l-cyclopentylglycine | |
CC(C)(C)OC(=O)NC(C1CCCC1)C(=O)O | |
243.303 | |
424.62 | |
1 g | |
N-Boc-2-cyclopentyl-L-glycine dicyclohexylammonium salt |
White | |
MFCD05663437 | |
WBSJQVRMQOLSAT-VIFPVBQESA-N | |
(2S)-2-cyclopentyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]acetic acid | |
7020746 | |
95% | |
Powder |
Safety and Handling
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C