missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-2-cyclopentyl-L-glycine dicyclohexylammonium salt, 95%

Manufacturer:  Alfa Aesar™ H62838MD

 View more versions of this product

Catalog No. AAH62838MD


 

Specifications

Specifications

N-Boc-2-cyclopentyl-L-glycine dicyclohexylammonium salt
95%
MFCD05663437
WBSJQVRMQOLSAT-VIFPVBQESA-N
(2S)-2-cyclopentyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]acetic acid
7020746
Powder
250mg
109183-72-4
C12H21NO4
Boc-Cpg-OH.DCHA
CC(C)(C)OC(=O)NC(C1CCCC1)C(=O)O
243.303
424.62
White

Chemical Identifiers

109183-72-4
243.303
WBSJQVRMQOLSAT-VIFPVBQESA-N
7020746
C12H21NO4
MFCD05663437
Boc-Cpg-OH.DCHA
Safety and Handling

Safety and Handling

storageNote1 : -30° to -10°C

SDS Documents