Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-2-cyclohexyl-L-glycine, 97%

Chemical Identifiers

Chemical Name or Material N-Boc-2-cyclohexyl-L-glycine
CAS 109183-71-3
Formula Weight 257.33
MDL Number MFCD00133638
Beilstein 5553687
Synonym Boc-Chg-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6246603

 
alfa aesar™
H6246603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6246606

 
alfa aesar™
H6246606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 257.33
Chemical Name or Material N-Boc-2-cyclohexyl-L-glycine
Physical Form Solid
Assay 97%
MDL Number MFCD00133638
CAS 109183-71-3
Beilstein 5553687
Synonym Boc-Chg-OH
Quantity 1g

Applications
N-Boc-2-cyclohexyl-L-glycine is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.