missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-2-cyclohexyl-D-glycine, 95%

Manufacturer:  Alfa Aesar™ H6367303

 View more versions of this product

Catalog No. AAH6367303



Specifications

Specifications

N-Boc-2-cyclohexyl-D-glycine
C13H23NO4
7689661
QSUXZIPXYDQFCX-SNVBAGLBSA-N
(2R)-2-cyclohexyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]acetic acid
7014890
Solid
1g
70491-05-3
MFCD00133629
boc-d-chg-oh, boc-d-cyclohexylglycine, r-2-tert-butoxycarbonyl amino-2-cyclohexylacetic acid, boc-alpha-cyclohexyl-d-glycine, n-boc-2-cyclohexyl-d-glycine, 2r-n-tert-butoxycarbonyl-2-cyclohexylglycine, r-tert-butoxycarbonylamino-cyclohexyl-acetic acid, r-tert-butoxycarbonyl amino cyclohexyl acetic acid, 2r-tert-butoxycarbonyl amino cyclohexyl acetic acid, 2r-2-tert-butoxycarbonylamino-2-cyclohexyl-acetic acid
CC(C)(C)OC(=O)NC(C1CCCCC1)C(=O)O
257.33
257.33
95%

Chemical Identifiers

70491-05-3
257.33
QSUXZIPXYDQFCX-SNVBAGLBSA-N
7014890
CC(C)(C)OC(=O)NC(C1CCCCC1)C(=O)O
C13H23NO4
MFCD00133629
boc-d-chg-oh, boc-d-cyclohexylglycine, r-2-tert-butoxycarbonyl amino-2-cyclohexylacetic acid, boc-alpha-cyclohexyl-d-glycine, n-boc-2-cyclohexyl-d-glycine, 2r-n-tert-butoxycarbonyl-2-cyclohexylglycine, r-tert-butoxycarbonylamino-cyclohexyl-acetic acid, r-tert-butoxycarbonyl amino cyclohexyl acetic acid, 2r-tert-butoxycarbonyl amino cyclohexyl acetic acid, 2r-2-tert-butoxycarbonylamino-2-cyclohexyl-acetic acid
(2R)-2-cyclohexyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]acetic acid
Safety and Handling

Safety and Handling

storageNote1 : -30° to -10°C

SDS Documents