Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-2-cyclohexyl-D-glycine, 95%

Chemical Identifiers

Chemical Name or Material N-Boc-2-cyclohexyl-D-glycine
CAS 70491-05-3
Formula Weight 257.33
MDL Number MFCD00133629
Beilstein 7689661
Synonym Boc-D-Chg-OH; N-Boc-α-cyclohexyl-D-glycine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6367303

  SDS
alfa aesar™
H6367303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6367306

  SDS
alfa aesar™
H6367306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 257.33
Physical Form Solid
Beilstein 7689661
MDL Number MFCD00133629
CAS 70491-05-3
Quantity 1g
Storage Note 1 -30° to -10°C
Assay 95%
Synonym Boc-D-Chg-OH; N-Boc-α-cyclohexyl-D-glycine
Chemical Name or Material N-Boc-2-cyclohexyl-D-glycine
Safety and Handling

-30° to -10°C