missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-2-cyano-D-phenylalanine, 95%

$114.43 - $342.35

Chemical Identifiers

CAS 261380-28-3
Molecular Formula C15H18N2O4
Molecular Weight (g/mol) 290.319
MDL Number MFCD01862940
InChI Key AJHPGXZOIAYYDW-GFCCVEGCSA-N
Synonym 2-cyano-n-2-methyl-2-propanyl oxy carbonyl-d-phenylalanine
PubChem CID 7010029
IUPAC Name (2R)-3-(2-cyanophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NC(CC1=CC=CC=C1C#N)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH62452MD View Documents Alfa Aesar™
H62452MD
250mg Each for $114.43
 
AAH6245203 View Documents Alfa Aesar™
H6245203
1g Each for $342.35
 
Specifications

Chemical Identifiers

261380-28-3
290.319
AJHPGXZOIAYYDW-GFCCVEGCSA-N
7010029
CC(C)(C)OC(=O)NC(CC1=CC=CC=C1C#N)C(=O)O
C15H18N2O4
MFCD01862940
2-cyano-n-2-methyl-2-propanyl oxy carbonyl-d-phenylalanine
(2R)-3-(2-cyanophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid

Specifications

261380-28-3
MFCD01862940
2-cyano-n-2-methyl-2-propanyl oxy carbonyl-d-phenylalanine
CC(C)(C)OC(=O)NC(CC1=CC=CC=C1C#N)C(=O)O
290.319
290.31
95%
250mg
C15H18N2O4
UN3439
AJHPGXZOIAYYDW-GFCCVEGCSA-N
(2R)-3-(2-cyanophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
7010029
Powder
White
N-Boc-2-cyano-D-phenylalanine
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
P261-P280-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.