missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-2-cyano-D-phenylalanine, 95%

Manufacturer:  Alfa Aesar™ H6245203

 View more versions of this product

Catalog No. AAH6245203



Specifications

Specifications

N-Boc-2-cyano-D-phenylalanine
C15H18N2O4
UN3439
AJHPGXZOIAYYDW-GFCCVEGCSA-N
(2R)-3-(2-cyanophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
7010029
Powder
White
261380-28-3
MFCD01862940
boc-2-cyano-d-phenylalanine, boc-d-2-cyanophenylalanine, r-2-tert-butoxycarbonyl amino-3-2-cyanophenyl propanoic acid, boc-d-phe 2-cn-oh, 2r-2-tert-butoxycarbonyl amino-3-2-cyanophenyl propanoic acid, boc-d-2-cyanophe, boc-d-2-cyano phenylalanine, boc-d-phe 2-cn-oh hplc, d-phenylalanine, 2-cyano-n-1,1-dimethylethoxy ca, 2-cyano-n-2-methyl-2-propanyl oxy carbonyl-d-phenylalanine
CC(C)(C)OC(=O)NC(CC1=CC=CC=C1C#N)C(=O)O
290.319
290.31
95%
1g

Chemical Identifiers

261380-28-3
290.319
AJHPGXZOIAYYDW-GFCCVEGCSA-N
7010029
CC(C)(C)OC(=O)NC(CC1=CC=CC=C1C#N)C(=O)O
C15H18N2O4
MFCD01862940
boc-2-cyano-d-phenylalanine, boc-d-2-cyanophenylalanine, r-2-tert-butoxycarbonyl amino-3-2-cyanophenyl propanoic acid, boc-d-phe 2-cn-oh, 2r-2-tert-butoxycarbonyl amino-3-2-cyanophenyl propanoic acid, boc-d-2-cyanophe, boc-d-2-cyano phenylalanine, boc-d-phe 2-cn-oh hplc, d-phenylalanine, 2-cyano-n-1,1-dimethylethoxy ca, 2-cyano-n-2-methyl-2-propanyl oxy carbonyl-d-phenylalanine
(2R)-3-(2-cyanophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
P261-P280-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

Documents