Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-2-chloro-D-phenylalanine, 95%

Chemical Identifiers

Chemical Name or Material N-Boc-2-chloro-D-phenylalanine
CAS 80102-23-4
Formula Weight 299.75
MDL Number MFCD00801056
Beilstein 8422927
Synonym Boc-D-Phe(2-Cl)-OH; Boc-D-2-chlorophe
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6110703

  SDS
alfa aesar™
H6110703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6110706

  SDS
alfa aesar™
H6110706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 299.75
Melting Point 94° to 98°C
CAS 80102-23-4
Assay 95%
Synonym Boc-D-Phe(2-Cl)-OH; Boc-D-2-chlorophe
MDL Number MFCD00801056
Chemical Name or Material N-Boc-2-chloro-D-phenylalanine
Beilstein 8422927
Physical Form Solid
Quantity 1g