Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-2-bromobenzylamine, 97%

Chemical Identifiers

Chemical Name or Material N-Boc-2-bromobenzylamine
CAS 162356-90-3
Formula Weight 286.17
MDL Number MFCD05664385
UN Number UN3077
Synonym N-(tert-Butoxycarbonyl)-2-bromobenzylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5519003

  SDS
alfa aesar™
H5519003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5519006

  SDS
alfa aesar™
H5519006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH55190MD

  SDS
alfa aesar™
H55190MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 286.17
Melting Point 50° to 54°C
MDL Number MFCD05664385
Color White to brown
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
DOT Information Transport Hazard Class: 9; Packaging Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
Chemical Name or Material N-Boc-2-bromobenzylamine
Physical Form Solid
Assay 97%
CAS 162356-90-3
UN Number UN3077
Quantity 1g
Synonym N-(tert-Butoxycarbonyl)-2-bromobenzylamine
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.