missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-2-bromobenzylamine, 97%

Manufacturer:  Alfa Aesar™ H5519006

 View more versions of this product

Catalog No. AAH5519006



Specifications

Specifications

N-Boc-2-bromobenzylamine
White to Brown
5g
C12H16BrNO2
UN3077
DFNZFCPEUDSNEO-UHFFFAOYSA-N
tert-butyl N-[(2-bromophenyl)methyl]carbamate
7018300
Solid
>110°C (230°F)
50°C to 54°C
162356-90-3
MFCD05664385
1-tert-butoxycarbonylaminomethyl-2-bromobenzene
CC(C)(C)OC(=O)NCC1=CC=CC=C1Br
286.169
286.17
97%

Chemical Identifiers

162356-90-3
286.169
DFNZFCPEUDSNEO-UHFFFAOYSA-N
7018300
CC(C)(C)OC(=O)NCC1=CC=CC=C1Br
C12H16BrNO2
MFCD05664385
1-tert-butoxycarbonylaminomethyl-2-bromobenzene
tert-butyl N-[(2-bromophenyl)methyl]carbamate
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

DOTInformation : Transport Hazard Class: 9; Packaging Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

Documents