Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-2-bromo-D-phenylalanine, 95%

Chemical Identifiers

Chemical Name or Material N-Boc-2-bromo-D-phenylalanine
CAS 261360-76-3
Formula Weight 344.2
MDL Number MFCD01317703
Synonym (R)-N-Boc-2-bromophenylalanine; Boc-Phe(2-Br)-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6284806

 
alfa aesar™
H6284806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6284803

 
alfa aesar™
H6284803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH62848MD

 
alfa aesar™
H62848MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 344.2
Chemical Name or Material N-Boc-2-bromo-D-phenylalanine
Physical Form Powder
Assay 95%
MDL Number MFCD01317703
CAS 261360-76-3
Synonym (R)-N-Boc-2-bromophenylalanine; Boc-Phe(2-Br)-OH
Color White
Quantity 5g

Applications
N-Boc-2-bromo-D-phenylalanine is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Notes
Keep container tightly closed. Store away from oxidizing agent.