Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-2-aminoisobutyric acid, 98+%

$27.57 - $56.36

Chemical Identifiers

Chemical Name or Material N-Boc-2-aminoisobutyric acid
CAS 30992-29-1
Molecular Formula C9H17NO4
Formula Weight 203.24
MDL Number MFCD00042973
Beilstein 1953772
Synonym Boc-Aib-OH; N-tert-Butoxycarbonyl-2-aminoisobutyric acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1948403

  SDS
alfa aesar™
L1948403
1g Each for $27.57

AAL1948406

  SDS
alfa aesar™
L1948406
5g Each for $56.36
Description & Specifications

Specifications

Formula Weight 203.24
Chemical Name or Material N-Boc-2-aminoisobutyric acid
MDL Number MFCD00042973
CAS 30992-29-1
Assay ≥98%
Molecular Formula C9H17NO4
Beilstein 1953772
Melting Point 118° to 121°C
Quantity 1g
Synonym Boc-Aib-OH; N-tert-Butoxycarbonyl-2-aminoisobutyric acid