Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-2,4,5-trifluoro-D-phenylalanine, 98%

Chemical Identifiers

Chemical Name or Material N-Boc-2,4,5-trifluoro-D-phenylalanine
CAS 486460-09-7
Formula Weight 319.28
MDL Number MFCD06659118
Synonym (R)-2-(Boc-amino)-3-(2,4,5-trifluorophenyl)propionic acid; Boc-D-2,4,5-trifluorophe
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6287803

 
alfa aesar™
H6287803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6287806

 
alfa aesar™
H6287806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH62878MD

 
alfa aesar™
H62878MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 319.28
MDL Number MFCD06659118
CAS 486460-09-7
Assay 98%
Physical Form Solid
Quantity 1g
Chemical Name or Material N-Boc-2,4,5-trifluoro-D-phenylalanine
Synonym (R)-2-(Boc-amino)-3-(2,4,5-trifluorophenyl)propionic acid; Boc-D-2,4,5-trifluorophe