Learn More
N-Boc-2-(3-pyridyl)-D-alanine, 95%, Thermo Scientific™
$92.73 - $240.33
Chemical Identifiers
CAS | 98266-33-2 |
---|---|
Molecular Formula | C13H18N2O4 |
Molecular Weight (g/mol) | 266.297 |
MDL Number | MFCD00076901 |
InChI Key | JLBCSWWZSSVXRQ-SNVBAGLBSA-N |
Synonym | boc-d-3-pyridylalanine, boc-d-3-pal-oh, boc-3-3-pyridyl-d-alanine, r-2-tert-butoxycarbonyl amino-3-pyridin-3-yl propanoic acid, r-n-boc-3-pyridyl alanine, boc-d-3-3-pyridyl-alanine, n-tert-butoxycarbonyl-3-pyridyl-d-alanine, boc-3'-pyridyl-d-ala, boc-3-3-pyridyl-d-ala-oh, boc-3-3'-pyridyl-d-alanine |
PubChem CID | 2734500 |
IUPAC Name | (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-pyridin-3-ylpropanoic acid |
SMILES | CC(C)(C)OC(=O)NC(CC1=CN=CC=C1)C(=O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH51985MD
|
Thermo Scientific Chemicals
H51985MD |
250 mg |
Each for $92.73
|
|
|||||
AAH5198503
|
Thermo Scientific Chemicals
H5198503 |
1 g |
Each for $240.33
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
98266-33-2 | |
266.297 | |
JLBCSWWZSSVXRQ-SNVBAGLBSA-N | |
2734500 | |
CC(C)(C)OC(=O)NC(CC1=CN=CC=C1)C(=O)O |
C13H18N2O4 | |
MFCD00076901 | |
boc-d-3-pyridylalanine, boc-d-3-pal-oh, boc-3-3-pyridyl-d-alanine, r-2-tert-butoxycarbonyl amino-3-pyridin-3-yl propanoic acid, r-n-boc-3-pyridyl alanine, boc-d-3-3-pyridyl-alanine, n-tert-butoxycarbonyl-3-pyridyl-d-alanine, boc-3'-pyridyl-d-ala, boc-3-3-pyridyl-d-ala-oh, boc-3-3'-pyridyl-d-alanine | |
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-pyridin-3-ylpropanoic acid |
Specifications
98266-33-2 | |
MFCD00076901 | |
boc-d-3-pyridylalanine, boc-d-3-pal-oh, boc-3-3-pyridyl-d-alanine, r-2-tert-butoxycarbonyl amino-3-pyridin-3-yl propanoic acid, r-n-boc-3-pyridyl alanine, boc-d-3-3-pyridyl-alanine, n-tert-butoxycarbonyl-3-pyridyl-d-alanine, boc-3'-pyridyl-d-ala, boc-3-3-pyridyl-d-ala-oh, boc-3-3'-pyridyl-d-alanine | |
CC(C)(C)OC(=O)NC(CC1=CN=CC=C1)C(=O)O | |
266.297 | |
266.3 | |
250 mg |
C13H18N2O4 | |
4195466 | |
JLBCSWWZSSVXRQ-SNVBAGLBSA-N | |
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-pyridin-3-ylpropanoic acid | |
2734500 | |
95% | |
N-Boc-2-(3-pyridyl)-D-alanine |
Safety and Handling
GHS H Statement
H317
May cause an allergic skin reaction.
GHS P Statement
P261-P280-P363-P321-P333+P313-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Specific treatment (see label).
If skin irritation or rash occurs: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
TSCA : No
Recommended Storage : Ambient temperatures