missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-1H-pyrazole-1-carboxamidine, 98+%

Manufacturer:  Alfa Aesar™ H5439403

 View more versions of this product

Catalog No. AAH5439403



Specifications

Specifications

N-Boc-1H-pyrazole-1-carboxamidine
1g
C9H14N4O2
6141956
IGSFMHYSWZUENI-UHFFFAOYSA-N
tert-butyl (NE)-N-[amino(pyrazol-1-yl)methylidene]carbamate
9605068
≥98%
97°C to 101°C
152120-61-1
MFCD00216663
1h-pyrazole-n-t-butyloxycarbonyl-1-carboxamidine
CC(C)(C)OC(=O)N=C(N)N1C=CC=N1
210.237
210.23

Chemical Identifiers

152120-61-1
210.237
IGSFMHYSWZUENI-UHFFFAOYSA-N
9605068
CC(C)(C)OC(=O)N=C(N)N1C=CC=N1
C9H14N4O2
MFCD00216663
1h-pyrazole-n-t-butyloxycarbonyl-1-carboxamidine
tert-butyl (NE)-N-[amino(pyrazol-1-yl)methylidene]carbamate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents