Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-1-(p-toluenesulfonyl)-L-histidine dicyclohexylammonium salt, 98%

Chemical Identifiers

Chemical Name or Material N-Boc-1-(p-toluenesulfonyl)-L-histidine dicyclohexylammonium salt
CAS 65057-34-3
Formula Weight 590.77
MDL Number MFCD00039005
Synonym Boc-His(Tos)-OH.DCHA; N-Boc-1-(p-tosyl)-L-histidine dicyclohexylammonium salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6220506

 
alfa aesar™
H6220506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6220514

 
alfa aesar™
H6220514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 590.77
Chemical Name or Material N-Boc-1-(p-toluenesulfonyl)-L-histidine dicyclohexylammonium salt
Physical Form Solid
Assay 98%
MDL Number MFCD00039005
CAS 65057-34-3
Synonym Boc-His(Tos)-OH.DCHA; N-Boc-1-(p-tosyl)-L-histidine dicyclohexylammonium salt
Quantity 5g